Transfection reagent, in vivo SilenceMag™
Lieferant: OZ BIOSCIENCESGesamtbewertung: 0
Bewertung: von 5
In vivo SilenceMag™ is a rapid, simple and highly efficient method dedicated to transfection of small RNA (siRNA, miRNA) into target cells/tissue in vivo. It combines magnetic nanoparticles and small RNA that will be retained after injection at the magnetically targeted site. This targeted delivery method minimises systemic distribution, increases gene targeted inactivation and reduces toxicity. Furthermore, the magnetic forces enhance the uptake of magnetic nanoparticles by the target tissue, and thus improve the efficiency of silencing.
- Increased silencing efficiency
- Targeted process (magnetically-driven)
- Reduction of the systemic dissemination of siRNA/miRNA during injection
- Reduction of the siRNA/miRNA doses
- Work under non permissive conditions (hypothermia, physiological flow conditions)
- Penetration of the siRNA/miRNA into tissues
- Minimum toxicity
Store at +4 °C.
Zubehörinformationen: This reagent needs to be used with a magnetic plate.
Lieferumfang: Shipped at room temperature.