Drucken
Transduction enhancer, AdenoMag™
Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
AM70100 AM70200 AM71000 KC30900
OZBIAM70100EA 118.25 EUR
OZBIAM70100 OZBIAM70200 OZBIAM71000 OZBIKC30900
Transduction enhancer, AdenoMag™
Transfektionsreagenzien
AdenoMag™ transfection reagent is a magnetic nanoparticles formulation specifically dedicated to enhance Adenovirus and Adeno Associated Virus (AAV) infection and transduction capacities. Based on Magnetofection™ technology, it allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
  • Increase transduction efficiency
  • Accelerate the transduction process
  • Efficient for hard-to-infect and non permissive cells
  • Concentrate the viral dose onto the cells
  • Synchronise cells adsorption/infection without modifications of the viruses
AdenoMag™ reagent demonstrates an exceptionally high efficiency to promote, control and assist Adenoviral and AAV transductions in such a way that no molecular biology processes or biochemical modifications are required.
Store at +4 °C.
Bestellinformation: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Lieferumfang: Shipped at room temperature.