DNA transfection reagent, PolyMag™
Lieferant: OZ BIOSCIENCESGesamtbewertung: 0
Bewertung: von 5
PolyMag™ transfection reagent, based on Magnetofection™ transfection technology, has been optimised for use with all types of nucleic acids. The reagent is composed of magnetic nanoparticles coated with cationic molecules. It has been successfully tested on a large range of primary cells, hard to transfect cells and cell lines and its efficiency has been reported in many publications.
- Highly efficient for primary cells
- Powerful for hard to transfect cells
- Non toxic
- Serum-compatible
- Simple, rapid and easy to use
- For all types of nucleic acids
Store at +4°C.
Zubehörinformationen: This reagent needs to be used with a magnetic plate.
Lieferumfang: Shipped at room temperature.