siRNA transfection reagent, SilenceMag™
Lieferant: OZ BIOSCIENCESGesamtbewertung: 0
Bewertung: von 5
SilenceMag™ siRNA delivery reagent is based on Magnetofection™ technology. Specifically designed for siRNA transfection, SilenceMag™ gives reliable high protein knockdown at very low doses of siRNA in numerous cell types (primary cells, hard-to-transfect and cell lines).
- High protein knockdown efficiency
- For all siRNA applications
- No off-target effects
- Serum compatible and non toxic
- Simple, rapid, and easy to use
Store at +4 °C.
Zubehörinformationen: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Lieferumfang: Shipped at room temperature.