Drucken
DNA transfection reagent, 3D-FectIN™
Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
TN30250 TN30500 TN31000
OZBITN30250EA 126.85 EUR
OZBITN30250 OZBITN30500 OZBITN31000
DNA transfection reagent, 3D-FectIN™
Transfektionsreagenzien
3D-FectIN™ is a 3D-transfection reagent specifically developed to directly transfect cells cultured in 3D hydrogels.
  • Highly efficient for cell lines and primary cells
  • Specific for 3D hydrogels
  • Compatible with all types of nucleic acids
  • Longterm protein expression
  • Serum compatible
Suitable for all type of nucleic acids including plasmid DNA, linearised DNA, double stranded RNA, mRNA, shRNA, oligonucleotides.
3D-FectIN™/DNA complexes are colonised by cells which are subsequently transfected in situ and in a more natural environment. This method allows studying angiogenesis, tube and acini formation, colonisation, neurite growth, tissue engineering, tissue regeneration, tumour invasion, neural differentiation, cellular polarisation and tissue formation.
Lieferumfang: Shipped at room temperature.
Achtung! Bitte beachten: Store at 4 °C.