Drucken

X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid) substrate 99, Molecular Biology Grade

Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

XG31000
OZBIXG31000EA 120.4 EUR
OZBIXG31000
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid) substrate 99, Molecular Biology Grade
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)

X-Gal substrate is metabolised by the b-galactosidase enzyme. The quality and purity of the substrate is essential to obtain high-quality and reliable results.


  • High purity molecular biology grade
  • Great sensitivity for cell and tissue staining


Designed for staining transformed bacteria and LacZ transfected or infected cells, tissues and organisms.


Lieferumfang: Shipped at room temperature.

Formel: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
MDL Number: MFCD00005666
CAS-Nummer: 7240-90-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9518ead23eb79069011509f486b02ef7d792eb46&vid=9518ead23eb79069011509f486b02ef7d792eb46&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR