Drucken
Percoll®, Siam
Lieferant: SIGMA ALDRICH MICROSCOPY
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
P1644-1L P1644-25ML P1644-500ML P1644-100ML
P1644-1LEA 939 EUR
P1644-1L P1644-25ML P1644-500ML P1644-100ML
Percoll®, Siam
Zelltrennmedien
Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particles of 15 – 30 nm diameter (23% w/w in water), coated with polyvinylpyrrolidone (PVP).
  • Non toxic
  • Ideal for use with biological materials