Drucken
Cleanroom ESD Ergo Chairs
Lieferant: Palbam
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
PCKD0100 PCKD0110 PCKD0120
PALBPCKD0100EA 1780 EUR
PALBPCKD0100 PALBPCKD0110 PALBPCKD0120
Cleanroom ESD Ergo Chairs
Stühle
Cleanroom ESD ergo chair which lives up to our reputation for high quality and ergonomics/operator comfort.
  • Seat and backrest material and colour: Polyurethane (PU), Black (RAL 9011)
  • Base Material: Chrome steel
Seat height adjustable from 450 to 640 mm.
The chairs include the 'automatic weight adjustment' ergo feature and a tapering backrest offering optimum freedom for shoulders. Options available for conductive glides or casters.
Zertifizierungen: The chairs comply with: Cleanliness class 4 DIN EN ISO 14644-1 Anti-static and conductive according to DIN EN 61340-5-1:2000.