Drucken
Trace metals (dissolved), Proficiency Testing Materials, Supelco®
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
PE1005-20ML
PE1005-20MLEA 94.6 EUR
PE1005-20ML
Trace metals (dissolved), Proficiency Testing Materials, Supelco®
Standards Kationischer Standards Mehrelement-Standards
Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.
Zertifizierungen: This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010.