Drucken

Sulphide (total and soluble) - WP, concentrate for dilution to 1 L, proficiency testing material, Supelco®

Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PE1034-20ML
PE1034-20MLEA 93.2 EUR
PE1034-20ML
Sulphide (total and soluble) - WP, concentrate for dilution to 1 L, proficiency testing material, Supelco®
Standards Anionische Standards Sulfid-Standards

This PT sample is as a 20 ml concentrate.


  • All constituents are present in soluble form
  • The solvent for this sample is water


The PT sample has been autoclaved for preservation.


Zertifizierungen: This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR