MALDI calibration kit, SpheriCal™ Aqua, peptide high, for MALDI MS, Supelco®
Gesamtbewertung: 0
Bewertung: von 5
SpheriCal™ is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal™ helps users increase the frequency and quality of their MALDI-MS calibrations.
- Mol wt.: Mr 2500-5000 Da
With a shelf life of years and each kit offering at least 100 calibrations, SpheriCal™ offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems
SpheriCal™ Aqua is ideal for calibration with aqueous systems commonly used in life sciences, e.g., matrices DHB, HCCA and SA or solvents such as water and acetonitrile, although ionization is preferred with alkali cations. Each vial of SpheriCal™ Neat contains sufficient calibration standard for over 100 calibration spots.
Achtung! Bitte beachten: SpheriCal is a trademark of Polymer Factory Sweden AB.