Drucken
Immobilisation kit for peptides, SulfoLink™, Pierce™
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
44999
PIER44999EA 461 EUR
PIER44999
Immobilisation kit for peptides, SulfoLink™, Pierce™
Protein-Aufbereitungs Protein Purification Kits
Immobilisation Kit for Peptides, SulfoLink™ is a kit containing all the reagents for preparing the peptide sample for efficient immobilisation and columns containing SulfoLink™ resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification.
  • Specific to sulphhydryls (-SH) groups
  • Prepare and couple protein samples in two hours
  • Flexible coupling conditions, use pH 7,5 to 9,0 aqueous buffers, organic solvent or denaturant
  • Streamlined protocols for sample preparation, immobilisation, and affinity purification
  • Immobilise 1 to 2 mg of peptide per 2 ml column of SulfoLink™ Coupling Resin
When incubated with a solution of peptide that contains reduced cysteine residues, the iodoacetyl groups of SulfoLink™ Resin react specifically and efficiently with the exposed sulphhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the peptide to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.
Lieferumfang: Kit include SulfoLink™ columns, 2 ml (5 columns), SulfoLink™ coupling buffer (120 ml), wash solution (120 ml), PBS pack (makes 500 ml), Bond-Breaker TCEP solution, neutral pH (0,5 ml), L-Cysteine·HCl (100 mg), and column accessories.