Affinity purification kits, Melon™ gel Monoclonal IgG purification kit
Lieferant: Thermo ScientificGesamtbewertung: 0
Bewertung: von 5
Melon™ Gel Monoclonal IgG Purification Kit is a high-yield, mild purification system for monoclonal antibodies from ascites or hybridoma cell culture supernatant, or for large scale antibody purification from serum.
- Higher recovery and purity
- Effective for subclasses of IgG that bind poorly to Protein A or G
- Regenerable, support can be regenerated and used for up to 6 purifications
- Compatible, amine free buffer does not require removal or neutralisation
Lieferumfang: Kit includes Melon™ gel support, 200 ml; Melon™ gel purification buffer (dry mix to make 100X), 1 L; and Melon™ gel regenerant (dry mix to make 5X), 1 L.