Drucken
Pierce™ Protein A/G Coated Plate IP Kit
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
45350
PIER45350EA 504 EUR
PIER45350
Pierce™ Protein A/G Coated Plate IP Kit
Protein-Aufbereitungs Protein Purification Kits
Pierce™ protein A/G coated plate IP kit uses coated protein A/G microplates to perform immunoprecipitation assays in 96-well plates without resins, beads, centrifugation or magnets.
  • Ready to use, high quality coated plates provide high capacity and consistency
  • Plate format best suited for simultaneously processing multiple samples and their control conditions
  • Faster, easier and more thorough washing than with traditional tube/resin IP methods
Lieferumfang: Protein A/G coated 96-well strip plates (2 plates), uncoated 96-well strip plate, white (2 plates), PBS pack (makes 500 ml), Surfact-Amps™ X-100 detergent solution (6×10 ml), elution buffer pH 2,8 (50 ml), neutralisation buffer (12 ml), and sealing tape for 96-well plates (18 sheets).