Drucken
Pierce™ DNA Extraction Reagent Kit, Yeast
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
78870
PIER78870EA 247 EUR
PIER78870
Pierce™ DNA Extraction Reagent Kit, Yeast
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The Pierce™ yeast DNA extraction reagent kit uses Y-PER yeast protein extraction reagent to quickly extract and purify genomic and plasmid DNA from yeast.
  • Greater yields than traditional methods
  • Eliminates need for glass beads or harsh enzyme treatments
  • Yields DNA that is compatible with PCR amplification
  • Scalable to allow purification from single colonies up to 500 ml of cultured yeast
Lieferumfang: Kit includes Y-PER yeast protein extraction reagent (25 ml), DNA releasing Agent A and B (20 ml of each), and protein removal reagent (10 ml).