Drucken
Western blot roller, Pierce™
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
84747
PIER84747EA 58.25 EUR
PIER84747
Western blot roller, Pierce™
Blotting-Apparate
The Western blot roller is a simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.
  • Reusable
  • Durable
The transfer-stack roller has a stainless steel handle and an inert PTFE roller that is 8,4 cm wide. Mini-gel sandwiches can be rolled out with one or two passes, but the tool also works well for transfer stacks with larger gels.
Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures.