Drucken
Anti-c-Myc magnetic beads, Pierce™
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
88842 88843
PIER88842EA 477 EUR
PIER88842 PIER88843
Anti-c-Myc magnetic beads, Pierce™
Kugeln Magnetkugeln
The Pierce™ Anti-c-Myc magnetic beads are affinity particles for immunoprecipitation of recombinant c-Myc-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
  • Highly specific anti-c-Myc monoclonal antibody enables high yield and high purity immunoprecipitation
  • Fast and convenient with low non-specific binding
  • Beads are compatible with manual and automated workflows
Stable, pre-blocked beads and specific antibody minimize off-target binding for c-Myc-tag IP or co-IP experiments
Order Now
[{"stiboOrderingTable":"<!--Specifications Tab-->\r\n\t\t\t\t<h2>TECHNISCHE DATEN</h2>\r\n\t\t\t\t <div id="stiboSpecGrid">\r\n\t\t\t\t\t<table id="stiboSpecGridTable" width="100%" class="productGrid">\r\n\t\t\t\t\t\t<caption class="caption">\r\n\t\t\t\t\t\t</caption>\r\n\t\t\t\t\t\t<tbody>\r\n\t\t\t\t\t\t\t<tr>\r\n\t\t\t\t\t\t\t\t<th></th>\t\t\t\t\t\t\t\t\t\t\t\r\n\t\t\t\t\t\t\t</tr>\r\n\t\t\t\t\t\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Environmentally Preferable</td>\r\n\t\t\t<td class="productGrid bld" colspan="1"></td>\r\n\t\t\t</tr>\r\n\t\t<style type="text/css">\r\n \t.stibospecprodgrid {\r\n \t\t\tdisplay:block;\r\n \t\t\toverflow-x:auto;\r\n \t\t\t}\t\r\n</style>\r\n\r\n<script type="text/javascript">\r\n\r\n\t$(document).ready(function() {\r\n\t\tif (numCol("#stiboSpecGridTable") > 4){\r\n\t\t\t$('#stiboSpecGrid').addClass('stibospecprodgrid');\r\n\t\t}\t\t\r\n\t});\r\n\t\r\n\tfunction numCol(table) {\r\n\t var maxColNum = 0;\r\n\t var i=0;\r\n\t var trs = $(table).find("tr");\r\n\t for ( i=0; i<trs.length; i++ ) {\r\n\t maxColNum = Math.max(maxColNum, getColForTr(trs[i]));\r\n\t }\r\n\r\n\t return maxColNum;\r\n\t}\r\n\tfunction getColForTr(tr) {\r\n\r\n\t var tds = $(tr).find("td");\r\n\r\n\t var numCols = 0;\r\n\r\n\t var i=1;\r\n\t for ( i=1; i<tds.length; i++ ) {\r\n\t var span = $(tds[i]).attr("colspan");\r\n\r\n\t if ( span )\r\n\t numCols += parseInt(span);\r\n\t else {\r\n\t numCols++;\r\n\t }\r\n\t }\r\n\t return numCols;\r\n\t}\r\n</script>\t\t\t\t\t\t\t\t\r\n</tbody>\r\n\t\t\t\t\t</table>\r\n\t\t\t\t\t</div>"}]