Drucken
Organelle enrichment kits
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
89839
PIER89839EA 486 EUR
PIER89839
Organelle enrichment kits
Protein-Extraktion Protein Extraction Kits
Organelle enrichment kit for lysosomes enables enrichment of intact organelles from cells and tissue. The kit provides the media and reagents necessary to accomplish subcellular fractionation using high-speed density gradient centrifugation.
  • Efficient and easy to use - kit reagents and gradient centrifugation separate organelles from contaminating structures
  • Compatible - prepare samples for downstream applications, including 2D/MS, electron microscopy, disease profiling, gene expression, signal transduction and interaction or localisation studies
  • Complete - kits contain sufficient material for 25 applications
Subcellular fractionation simplifies otherwise complex protein mixtures that result from whole cell lysis, thereby facilitating proteomic analysis of specific organelle targets and pathways. Each kit uses density gradient centrifugation to separate organelles from contaminating cellular structures. The isolated organelles may be used for a number of downstream applications, including 2D/MS, electron microscopy, disease profiling, gene expression, signal transduction and interaction or localisation studies.
Lieferumfang: Each kit contains Enrichment Reagent A, 90 ml; Enrichment Reagent B, 90 ml; OptiPrep Cell Separation Media, 50 ml; PBS (makes 500 ml), 1 pack.