Drucken
In-Gel tryptic digestion kit, Pierce™
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
89871
PIER89871EA 352 EUR
PIER89871
In-Gel tryptic digestion kit, Pierce™
Protein-Extraktion Protein Extraction Kits
The Pierce™ In-Gel Tryptic Digestion Kit provides a complete set of reagents for digesting and recovering peptide fragments from approximately 150 stained protein bands excised from SDS-PAGE (polyacrylamide gel plugs).
  • Includes all necessary reagents for destaining Coomassie®- or fluorescent dye-stained proteins, reduction and alkylation of cystines, and tryptic digestion
  • Procedure and reagents produce reliable digestions and data generation using a wide range of conditions without optimisation
  • Contains highly purified and modified MS-grade trypsin that shows no chymotryptic activity and minimal autolytic activity
Lieferumfang: The kit includes modified porcine trypsin, destaining buffers, reduction reagents, alkylation reagents and digestion buffers along with detailed and simple instructions.