Drucken
Phosphoprotein enrichment kit, Pierce™
Lieferant: Thermo Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
90003
PIER90003EA 808 EUR
PIER90003
Phosphoprotein enrichment kit, Pierce™
Protein-Extraktion Protein Extraction Kits
Pierce™ Phosphoprotein Enrichment Kit is designed for the efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues.
  • Low non specific protein contamination from complex biological samples
  • Easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
  • High yields
Pierce™ Phosphoprotein Enrichment Kit uses a convenient spin column format and is compatible with downstream applications including Western blotting, mass spectronomy, 2D-PAGE, and protein arrays.