Phosphoprotein enrichment kit, Pierce™
Lieferant: Thermo ScientificGesamtbewertung: 0
Bewertung: von 5
Pierce™ Phosphoprotein Enrichment Kit is designed for the efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues.
- Low non specific protein contamination from complex biological samples
- Easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
- High yields
Pierce™ Phosphoprotein Enrichment Kit uses a convenient spin column format and is compatible with downstream applications including Western blotting, mass spectronomy, 2D-PAGE, and protein arrays.