Drucken

Anti-PDGFR beta(pTyr 751 ) Rabbit Polyclonal Antibody

Lieferant: Merck Millipore (Calbiochem‎)
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PK1008-50UL
PK1008-50ULEA 352 EUR
PK1008-50UL
Anti-PDGFR beta(pTyr 751 ) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDGFR beta(pTyr 751 )
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR