Drucken
STANDARD SEM PIN STUB MOUNT, FEI, TESCAN, ZEISS (ALSO PHILIPS, ASPEX, LEO, CAMBRIDGE, AMRAY, LEICA, CAMSCAN, ETEC), ALUMINIUM, GROOVED EDGE, D=12.7 MM X 8 MM 1 * 1.000 ST
Lieferant: PLANO W. PLANNET
STANDARD SEM PIN STUB MOUNT, FEI, TESCAN, ZEISS (ALSO PHILIPS, ASPEX, LEO, CAMBRIDGE, AMRAY, LEICA, CAMSCAN, ETEC), ALUMINIUM, GROOVED EDGE, D=12.7 MM X 8 MM 1 * 1.000 ST