Drucken
Tourniquets, easy
Lieferant: Praemeta
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
904
PRAE904EA 95.6 EUR
PRAE904
Tourniquets, easy
Stauschläuche
Tourniquets made of latex-free, skin-friendly, durable cotton fabric.
  • Suitable for fast blood sampling
  • High quality metal lock in a modern design
  • Separately and precisely adjustable decongestion
  • Reduced tissue acidosis during extended use
Suitable for doctor's bags and first-aid kits.