Drucken
Chlorate ion chromatography standards, VHG
Lieferant: PROMOCHEM
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
VHG-ICLO3-100 VHG-ICLO3-500
PROOVHG-ICLO3-100EA 58 EUR
PROOVHG-ICLO3-100 PROOVHG-ICLO3-500
Chlorate ion chromatography standards, VHG
Standards Anionische Standards Chlorat-Standards
This solution is intended for use as a certified reference material and calibration standard, for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Zertifizierungen: Traceable to NIST and CRM.