Drucken

Anti-MAP4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

19-937
PRSI19-937EA 419 EUR
PRSI19-937
Anti-MAP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR