Drucken

Anti-DHTKD1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

23-539
PRSI23-539EA 419 EUR
PRSI23-539
Anti-DHTKD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DHTKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR