Drucken

Anti-F2RL3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

23-606
PRSI23-606EA 419 EUR
PRSI23-606
Anti-F2RL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: F2RL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR