Drucken
Anti-GATA3 Rabbit Polyclonal Antibody
Lieferant: ProSci Inc.
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
25-031
PRSI25-031EA 660 EUR
PRSI25-031
Anti-GATA3 Rabbit Polyclonal Antibody
Antikörper
GATA3 is crucially involved in IL-5 gene transcription in human peripheral CD4-positive t cells. This gene is involved in growth control and the maintenance of the differentiated state in epithelial cells and may contribute to tumorigenesis in breast cancer.
GATA3 antibody can be used for detection of GATA3 by ELISA at 1:1562500. GATA3 antibody can be used for detection of GATA3 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS bufferMore Product Information