Drucken

Anti-ZNF146 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

25-386
PRSI25-386EA 660 EUR
PRSI25-386
Anti-ZNF146 Rabbit Polyclonal Antibody
Antikörper
ZNF146 (OZF) overexpression in tumours may alter the balance between hRap1 and other telomeric proteins; therefore OZF function may be linked to telomere regulation. ZNF146 is strongly overexpressed in many pancreas and colorectal cancers. Increased gene copy numbers are detected in 3 of 12 tumor cell lines and 2 of 12 primary pancreatic carcinomas. ZNF146 is overexpresseed in 80% of colorectal cancers.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZNF146
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR