Drucken
Anti-FOXR2 Rabbit Polyclonal Antibody
Lieferant: ProSci Inc.
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
29-806
PRSI29-806EA 601 EUR
PRSI29-806
Anti-FOXR2 Rabbit Polyclonal Antibody
Antikörper
Forkhead-box (FOX) transcription factors are implicated in carcinogenesis through gene amplification, retroviral integration, or chromosomal translocation. Human FOXN6 mRNA was expressed in breast cancer cell line and primary breast cancer.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FOXR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human