Drucken

Anti-PLIN2 Mouse Monoclonal Antibody [clone: ADPN1-1]

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

34-036
PRSI34-036EA 773 EUR
PRSI34-036
Anti-PLIN2 Mouse Monoclonal Antibody [clone: ADPN1-1]
Antikörper
Adipophilin belongs to the perilipin family, members of which coat intracellular lipid storage droplets. This protein is associated with the lipid globule surface membrane material and maybe involved in development and maintenance of adipose tissue. It is not restricted to adipocytes as previously thought, but is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells, and tissues, such as lactating mammary gland, adrenal cortex, Sertoli and Leydig cells, and hepatocytes in alcoholic liver cirrhosis, suggesting that it may serve as a marker of lipid accumulation in diverse cell types and diseases.

Flow Cytometry: 0,5 - 1 µg/10^6 cells Western blot: 1 - 2 µg/ml Titering of the Adipophilin antibody may be required for optimal performance.

Type: Primary
Antigen: PLIN2
Clonality: Monoclonal
Clone: ADPN1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR