Drucken

Anti-TP53 Mouse Monoclonal Antibody [clone: SPM590]

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

34-174
PRSI34-174EA 773 EUR
PRSI34-174
Anti-TP53 Mouse Monoclonal Antibody [clone: SPM590]
Antikörper
Recognises a 53 kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53. p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to MDM2, SV40 T antigen and human papilloma virus E6 protein. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia. Mutations involving p53 are found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,25-0,5 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the anti-p53 antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: SPM590
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Monkey, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR