Drucken

Anti-GALNT5 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

64-219
PRSI64-219EA 620 EUR
PRSI64-219
Anti-GALNT5 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: GALNT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 24-53
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR