Drucken

Anti-Ptprc Mouse Monoclonal Antibody [clone: A20] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

76-538
PRSI76-538EA 343 EUR
PRSI76-538
Anti-Ptprc Mouse Monoclonal Antibody [clone: A20] (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: A20
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR