Drucken

Anti-Ptprc Mouse Monoclonal Antibody [clone: 104] (APC (Allophycocyanin)-Cy7®)

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

76-565
PRSI76-565EA 522 EUR
PRSI76-565
Anti-Ptprc Mouse Monoclonal Antibody [clone: 104] (APC (Allophycocyanin)-Cy7®)
Antikörper
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 104
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR