Drucken

Anti-Cdh5 Rat Monoclonal Antibody [clone: BV13] (PE (Phycoerythrin))

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

76-864
PRSI76-864EA 343 EUR
PRSI76-864
Anti-Cdh5 Rat Monoclonal Antibody [clone: BV13] (PE (Phycoerythrin))
Antikörper
The BV13 monoclonal antibody specifically reacts with mouse CD144 (VE-Cadherin), a 120 kDa type II Cadherin. CD144 is a hemophilic adhesion molecule of the endothelial cell line essential to maintaining cell layer integrity. It is involved in cell growth inhibition, migration, and survival.

Type: Primary
Antigen: Cdh5
Clonality: Monoclonal
Clone: BV13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR