Drucken

Anti-LAMP1 Mouse Monoclonal Antibody [clone: H4A3] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

76-890
PRSI76-890EA 343 EUR
PRSI76-890
Anti-LAMP1 Mouse Monoclonal Antibody [clone: H4A3] (FITC (Fluorescein Isothiocyanate))
Antikörper
The H4A3 monoclonal antibody specifically reacts with human CD107a, a heavily glycosylated type I membrane glycoprotein. CD107a is also known as Lysosomal-associated membrane protein 1 (LAMP-1) and is widely expressed intracellular antigen. It can be found on the surface of PHA-activated lymphocytes, activated platelets, cytotoxic T cells, NK cells, macrophages, epithelial cells, endothelial cells, and some tumor lines. CD107a is a ligand for E-selectin and galaptin and is reported to be involved in cell adhesion and tumor metastasis.

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: H4A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR