Drucken

Anti-Klrg1 Hamster Monoclonal Antibody [clone: 2F1] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

77-091
PRSI77-091EA 448 EUR
PRSI77-091
Anti-Klrg1 Hamster Monoclonal Antibody [clone: 2F1] (FITC (Fluorescein Isothiocyanate))
Antikörper
The 2F1 monoclonal antibody is specific for the mouse Killer cell Lectin-like Receptor G1 (KLRG1), a homodimer consisting of two N-glycosylated subunits of 30-38 kDa, also known as MAFA (Mast cell Function-associated Antigen). The antigen contains a cytoplasmic motif similar to ITIM (the immunoreceptor tyrosine-based inhibitory motif). KLRG1 is a receptor for cadherin, a family of transmembrane glycoproteins that mediate cell adhesion, and a common marker of T cell senescence. The receptor is believed to play an important role in the innate and adaptive immune system through the regulation of leukocytes. It is expressed on lymphokine-activated killer (LAK) cells, adherent LAK (A-LAK) cells, a sub-set of natural killer (NK) cells, T cells. In NK cells, it inhibits cytokine production and cytotoxicity activity.The receptor expression was not detected on the mouse peritoneal mast cells, or bone marrow mast cells.

Type: Primary
Antigen: Klrg1
Clonality: Monoclonal
Clone: 2F1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR