Drucken
Anti-EGFR Rabbit Polyclonal Antibody
Lieferant: ProSci Inc.
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
79-208
PRSI79-208EA 469 EUR
PRSI79-208
Anti-EGFR Rabbit Polyclonal Antibody
Antikörper
Receptor for EGF, but also for other members of the EGF family, as TGF-alpha, amphiregulin, betacellulin, heparin-binding EGF-like growth factor, GP30 and vaccinia virus growth factor. Is involved in the control of cell growth and differentiation. Phosphorylates MUC1 in breast cancer cells and increases the interaction of MUC1 with SRC and CTNNB1/beta-catenin.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-More Product Information