Mouse recombinant Meteorin-like (from HEK293 cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90-330
PRSI90-330EA 448 EUR InStock
PRSI90-330
Mouse recombinant Meteorin-like (from HEK293 cells)
Proteine und Peptide
Meteorin-like (cometin; subfatin) is a potent new neurotrophic factor promoting neurite outgrowth from dorsal root ganglion cells. Meteorin-like is also a novel adipokine expressed by adipose tissue being downregulated upon caloric restriction. Meteorin-like is secreted by muscles during exercise and converts white adipose cells into brown fat tissue.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge