Human recombinant CXCL7 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-045
PRSI91-045EA 852 EUR InStock
PRSI91-045
Human recombinant CXCL7 (from E. coli)
Proteine und Peptide
Human Chemokine (C-X-C motif) Ligand 7 (CXCL7), also known as neutrophil activating peptide 2 (NAP-2), is a member of the CXC chemokines containing an ELR domain (Glu-Leu-Arg tripeptide motif). Similar to other ELR domain containing CXC chemokines, such as IL-8 and the GRO proteins, CXCL7 binds CXCR2, chemoattracts and activates neutrophils. CXCL7, Connective Tissue Activating Protein III (CTAPIII) and beta thrombogulin ( beta TG), are proteolytically processed carboxylterminal fragments of platelet basic protein (PBP) which is found in the alphagranules of human platelets. Although CTAPIII, beta TG, and PBP represent amino-terminal extended variants of NAP2 and possess the same CXC chemokine domains, these proteins do not exhibit CXCL7/NAP2 activity. CXCL7 induces cell migration through the G-protein-linked receptor CXCR-2.
  • This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge