Human recombinant Cystatin S (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-311
PRSI91-311EA 549 EUR InStock
PRSI91-311
Human recombinant Cystatin S (from cells)
Proteine und Peptide
Cystatin-S, also called Cystatin-SA-III or Salivary acidic protein, is usually expressed in submandibular and sublingual saliva but not in parotid saliva(at protein level). This protein, which is phosphorylated at both its N- and C-terminal regions, belongs to the cystatin family. Cystatin-S can function as an inhibitor to papain or ficin, it partially inhibits stem bromelain and bovine cathepsin C.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge