Human recombinant AGER (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-379
PRSI91-379EA 549 EUR InStock
PRSI91-379
Human recombinant AGER (from cells)
Proteine und Peptide
Advanced Glycosylation End Product-Specific Receptor (AGER) belongs to the immunoglobulin superfamily of cell surface molecules. It lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Besides AGEs, AGER is also able to bind other ligands which is thought to result in pro-inflammatory gene activation. It is known that AGER serve as a mediator of both acute and chronic vascular inflammation in certain conditions such as atherosclerosis and in particular as a complication of diabetes. Furthermore, it plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge