Human recombinant LILRB2 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-439
PRSI91-439EA 528 EUR InStock
PRSI91-439
Human recombinant LILRB2 (from cells)
Proteine und Peptide
Members of the immunoglobulin-like transcript (ILT) family are activating and inhibitory immunoreceptors whose genes are located same locus that encodes killer cell Ig-like receptors (KIR). Leukocyte Immunoglobulin-Like Receptor Subfamily B Member 2 (LIR-2) is a type I transmembrane protein. LIR-2 is expressed primarily on monocytes and dendritic cells (DC). Human LIR-2 is produced as a 598 amino acino acid precursor including a 21 aa signal sequence, a 440 aa extracellular domain (ECD), a 21 aa transmenbrane segment, and a 116 aa cytoplasmic domain. LIR-2 binds to Classical MHCI proteins. Ligation of LIR-2 incluces Tyr phosphorylation within its cytoplasmic ITIMs, a requirement for association with SHP-1. LIR-2 mediates tolerogenic DC-induced CD4+ T cell energy in vitro and in vivo.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge