Human recombinant PHLDA2 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-098
PRSI92-098EA 654 EUR InStock
PRSI92-098
Human recombinant PHLDA2 (from E. coli)
Proteine und Peptide
Pleckstrin Homology-Like Domain Family A Member 2 (PHLDA2) is a peripheral membrane protein that belongs to the PHLDA2 family. PHLDA2 is expressed in the placenta and adult prostate gland. In the placenta, it is present in all cells of the villous cytotrophoblast. PHLDA2 plays a role in regulating placenta growth. PHLDA2 may act via its PH domain that competes with other PH domain-containing proteins, thereby preventing their binding to membrane lipids.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge