Human recombinant STAT6 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-166
PRSI92-166EA 852 EUR InStock
PRSI92-166
Human recombinant STAT6 (from E. coli)
Proteine und Peptide
Signal Transducer and Activator of Transcription 6 (STAT6) is a member of the STAT family of transcription factors. At least seven STATs exist: STAT1, 2, 3, 4, 5a, 5b, and 6. They are responsible for an array of cellular activities including regulating growth, survival, differentiation, motility, and the immune response. STAT6 plays a central role in exerting IL4 mediated biological responses. It is found to induce the expression of BCL2L1/BCL-X(L), which is responsible for the anti-apoptotic activity of IL4. Knockout studies in mice suggested the roles of this gene in differentiation of T helper 2 (Th2) cells, expression of cell surface markers, and class switch of immunoglobulins. STAT6 has been shown to interact with EP300, CREB-binding protein, NFKB1, Nuclear receptor coactivator 1, IRF4 and SND1.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge