92-345
PRSI92-345EA
654
EUR
InStock
PRSI92-345
Human recombinant PFKM (from cells)
Proteine und Peptide
6-phosphofructokinase, muscle type is a muscle-type isozyme that in humans is encoded by the PFKM gene. It belongs to the phosphofructokinase family and Two domains subfamily. PFKM functions as subunits of the mammalian tetramer phosphofructokinase, which catalyses the phosphorylation of fructose-6-phosphate to fructose-1,6-bisphosphate. PFK1 converts fructose 6-phosphate and ATP into fructose 1,6-bisphosphate (through PFK-1), fructose 2,6-bisphosphate (through PFK-2) and ADP.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.