Human recombinant PFKM (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-345
PRSI92-345EA 654 EUR InStock
PRSI92-345
Human recombinant PFKM (from cells)
Proteine und Peptide
6-phosphofructokinase, muscle type is a muscle-type isozyme that in humans is encoded by the PFKM gene. It belongs to the phosphofructokinase family and Two domains subfamily. PFKM functions as subunits of the mammalian tetramer phosphofructokinase, which catalyses the phosphorylation of fructose-6-phosphate to fructose-1,6-bisphosphate. PFK1 converts fructose 6-phosphate and ATP into fructose 1,6-bisphosphate (through PFK-1), fructose 2,6-bisphosphate (through PFK-2) and ADP.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge