Drucken

Anti-Itga4 Rat Monoclonal Antibody [clone: PS/2] (PE (Phycoerythrin))

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

98-552
PRSI98-552EA 509 EUR
PRSI98-552
Anti-Itga4 Rat Monoclonal Antibody [clone: PS/2] (PE (Phycoerythrin))
Antikörper
Type: Primary
Antigen: Itga4
Clonality: Monoclonal
Clone: [PS/2]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR