Drucken

Anti-MDK Rabbit Polyclonal Antibody (Biotin)

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

XP-5230BT
PRSIXP-5230BTEA 509 EUR
PRSIXP-5230BT
Anti-MDK Rabbit Polyclonal Antibody (Biotin)
Antikörper
Midkine exhibits neurite outgrowth-promoting activity and may play a role in nervous system development and/or maintenance. Its expression is predominant only for a short period from approximately one-half to two-thirds of the way through gestation; before and after that, it is barely detectable. Midkine was first found in differentiating mouse teratocarcinoma cells. It has neurotrophic activities and is mitogenic to certain, but not to all, fibroblast cell lines.

Anti-hMidkine specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MDK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR