Drucken

Anti-CCL3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: ProSci Inc.
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

XP-5233BT
PRSIXP-5233BTEA 509 EUR
PRSIXP-5233BT
Anti-CCL3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Macrophage Inflammatory Protein 1 alpha and MIP1 beta, two closely related but distinct proteins, were originally co-purified from medium conditioned by a LPS-stimulated murine macrophage cell line. Mature mouse MIP1 alpha shares approximately 77% and 70% amino acid identity with human MIP1 alpha and mouse MIP1 beta, respectively. MIP1 proteins are expressed primarily in T cells, B cells, and monocytes after antigen or mitogen stimulation.

Anti-rat MIP-1-alpha specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR